5-Bromo-1-pentene ≥97%

Supplier: Matrix Scientific

034859-25G 034859-100G
101780-572EA 69.23 USD
101780-572 102551-164
5-Bromo-1-pentene ≥97%
5-Bromo-1-pentene

Hazard: Flammable

Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 126…127 °C
Density: 1.259
Flash Pt: 30 °C
MDL Number: MFCD00000264
CAS Number: 1119-51-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 126-127°
Density 1.259
ND20 1.4639

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR