3-Bromophenylhydrazinium chloride ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride
034730-25G
034730-100G
101780-556EA
66.15
USD
101780-556
101780-558
3-Bromophenylhydrazinium chloride ≥98%
3-Bromophenylhydrazinium chloride
Hazard: Corrosive/Irritant/Hygroscopic
Formula:
C₆H₈BrClN₂ Melting Pt: ca 225°C |
MDL Number:
MFCD00012933 CAS Number: 27246-81-7 |
Specification Test Results
Melting point | ca 225° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...