2,4-Difluoropyridine ≥98%
Supplier: Matrix Scientific
032593-5G
032593-25G
101780-548EA
75.38
USD
101780-548
102832-992
2,4-Difluoropyridine ≥98%
2,4-Difluoropyridine
Hazard: Flammable
Formula:
C₅H₃F₂N Boiling Pt: 106°C Density: 1.27 |
MDL Number:
MFCD04114162 CAS Number: 34941-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...