2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Fluoro-3-(trifluoromethyl)phenacyl bromide, 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone
032529-1G
032529-5G
101780-540EA
26.15
USD
101780-540
101780-542
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥95%
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
Hazard: Corrosive/Irritant-Harmful/Refrigerate/Lachrymator
Formula:
C₉H₅BrF₄O MW: 285.0330 g/mol Melting Pt: 42-44°C |
MDL Number:
MFCD03094509 CAS Number: 537050-14-9 |
Specification Test Results
Melting point | 42-44° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...