2,2-Dichloro-1,3-benzodioxole ≥95%
Supplier: Matrix Scientific
032431-1G
032431-5G
101780-534EA
75.38
USD
101780-534
101780-536
2,2-Dichloro-1,3-benzodioxole ≥95%
2,2-Dichloro-1,3-benzodioxole
Hazard: Flammable/Refrigerate
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol |
MDL Number:
MFCD00196818 CAS Number: 2032-75-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...