4-Bromo-3-methylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3-methylbenzene-1-sulphonyl chloride

032356-1G 032356-5G 032356-25G
101780-520EA 15.38 USD
101780-520 101780-522 102551-148
4-Bromo-3-methylbenzenesulfonyl chloride ≥97%
4-Bromo-3-methylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 57…61 °C
MDL Number: MFCD03094696
CAS Number: 72256-93-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 57-61°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR