Methyl-2-isocyanatobenzoate

Supplier: Matrix Scientific

Synonyms: 2-(Methoxycarbonyl)phenyl isocyanate

030179-5G 030179-25G
101780-502EA 181.54 USD
101780-502 101780-504
Methyl-2-isocyanatobenzoate
Methyl 2-isocyanatobenzoate

Hazard: Toxic

Formula: C₉H₇NO₃
MDL Number: MFCD00013848
CAS Number: 1793-07-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR