4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
026761-5G
026761-1G
101780-494EA
87.69
USD
101780-494
101780-492
4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride
Hazard: Corrosive/Irritant/Moisture Sensitive
Formula:
C₇H₃ClF₄O₂S Boiling Pt: 76 °C (0.5 mmHg) Melting Pt: 54…56 °C |
MDL Number:
MFCD01091000 CAS Number: 176225-09-5 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 76°/0.5mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...