4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

026756-1G 026756-5G
101780-484EA 15.38 USD
101780-484 101780-486
4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride ≥97%
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₃ClF₄O₂S
Boiling Pt: 92 °C (0.8 mmHg)
MDL Number: MFCD01091001
CAS Number: 1682-10-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 92°/0.8mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR