2,4-Dimethylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dimethylbenzene-1-sulphonyl chloride, m-Xylene-4-sulphonyl chloride

026307-5G 026307-100G 026307-25G
101780-472EA 83.08 USD
101780-472 101780-476 101780-474
2,4-Dimethylbenzenesulfonyl chloride ≥97%
2,4-Dimethylbenzenesulphonyl chloride

TCA listed: T
Hazard: Corrosive/Irritant/Moisture Sensitive

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Boiling Pt: 82°C/0.4mm
Melting Pt: 28-30°C
MDL Number: MFCD00156143
CAS Number: 609-60-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 82°/0.4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR