3-Oxopentanenitrile ≥97%

Supplier: Matrix Scientific

026306-1G 026306-5G
101780-466EA 56.3 USD
101780-466 101780-468 101780-470
3-Oxopentanenitrile ≥97%
3-Oxopentanenitrile

Hazard: Toxic/Store in Freezer

Formula: C₅H₇NO
MW: 97.117 g/mol
Boiling Pt: 30 °C
MDL Number: MFCD07367087
CAS Number: 33279-01-5

Order Now

Specification Test Results

Boiling point 30°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR