5-Fluoro-2-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

024845-1G 024845-5G
101780-462EA 60 USD
101780-462 101780-464
5-Fluoro-2-(trifluoromethyl)pyridine ≥98%
5-Fluoro-2-(trifluoromethyl)pyridine

Hazard: Flammable

Formula: C6H3F4N
MDL Number: MFCD09864698
CAS Number: 936841-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR