2-(Trimethylsilyl)ethanol ≥98%
Supplier: Matrix Scientific
024803-5G
024803-25G
024803-100G
101780-438EA
32.31
USD
101780-438
101780-440
102663-648
2-(Trimethylsilyl)ethanol ≥98%
2-(Trimethylsilyl)ethanol
Hazard: Flammable
Formula:
HOH₂CCH₂Si(CH₃)₃ Boiling Pt: 71-72°C/10mm Density: 0.825 |
MDL Number:
MFCD00002825 CAS Number: 2916-68-9 UN: 1987 ADR: 3,III |
Specification Test Results
Boiling point | 71-72°/10m |
Density | 0.825 |
ND20 | 1.425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...