2-Phthalimidoethanesulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)ethanesulphonylchloride

024771-1G 024771-5G
101780-432EA 29.23 USD
101780-432 101780-434
2-Phthalimidoethanesulfonyl chloride ≥95%
2-Phthalimidoethanesulfonyl chloride

Hazard: Corrosive/Irritant

Formula: C₁₀H₈ClNO₄S
MW: 273.7 g/mol
MDL Number: MFCD01861217
CAS Number: 4403-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR