3,5-Difluoropyridine ≥98%

Supplier: Matrix Scientific

024725-5G 024725-1G 024725-25G
101780-424EA 66.15 USD
101780-424 101780-422 101780-426
3,5-Difluoropyridine ≥98%
3,5-Difluoropyridine

Hazard: Flammable

Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 92-93°C
Density: 1.268
MDL Number: MFCD03412222
CAS Number: 71902-33-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 92-93°
Density 1.268
ND20 1.4437

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR