Benzothiazole-2-carbonyl chloride ≥95%
Supplier: Matrix Scientific
024699-1G
024699-250MG
101780-420EA
200
USD
101780-420
101780-418
Benzothiazole-2-carbonyl chloride ≥95%
Benzothiazole-2-carbonyl chloride
Hazard: Corrosive/Irritant
Formula:
C₈H₄ClNOS MW: 197.64 g/mol Melting Pt: 113…115 °C |
MDL Number:
MFCD03659697 CAS Number: 67748-61-2 |
Specification Test Results
Melting point | 113-115° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...