3-Fluoro-4-nitrobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-nitrobenzene-1-sulphonyl chloride

024659-1G 024659-5G
101780-412EA 75.38 USD
101780-412 102551-066
3-Fluoro-4-nitrobenzenesulfonyl chloride ≥98%
3-Fluoro-4-nitrobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₃ClFNO₄S
MW: 239.61 g/mol
Boiling Pt: 126°C/0.25mm
Melting Pt: 44-48°C
MDL Number: MFCD03412240
CAS Number: 86156-93-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 126°/0.25m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR