3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

024658-1G 024658-5G 024658-25G
101780-404EA 15.38 USD
101780-404 101780-406 101780-408
3-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive/Irritant

Formula: C₇H₃BrClF₃O₂S
Boiling Pt: 217…218 °C
MDL Number: MFCD03094393
CAS Number: 351003-46-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 217-218°
ND20 1.835


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR