3-Chloro-4-methylanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methoxy-1-methylbenzene, 3-Chloro-4-methylanisole, 2-Chloro-4-methoxytoluene

024623-5G 024623-25G
101780-392EA 36.92 USD
101780-392 101780-394
3-Chloro-4-methylanisole ≥97%
3-Chloro-4-methylanisole

Hazard: Flammable

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 76 °C (2 mmHg)
MDL Number: MFCD00087376
CAS Number: 54788-38-4

Order Now

Specification Test Results

Boiling point 76°/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR