4-Phenoxybenzenesulphonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Phenoxybenzene-1-sulphonyl chloride

024599-1G 024599-5G
101780-380EA 49.23 USD
101780-380 101780-382
4-Phenoxybenzenesulphonyl chloride ≥98%
4-Phenoxybenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₁₂H₉ClO₃S
MW: 268.72
Melting Pt: 44
MDL Number: MFCD00625748
CAS Number: 1623-92-3
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Melting point 43-44°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR