2,4,6-Trimethylphenylhydrazine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Mesitylhydrazine xhydrochloride

023814-1G 023814-5G
101780-352EA 80 USD
101780-352 101780-354
2,4,6-Trimethylphenylhydrazine hydrochloride ≥97%
2,4,6-Trimethylphenylhydrazine hydrochloride

Hazard: Toxic/Store in Freezer

Formula: C₉H₁₅ClN₂
MW: 186.69 g/mol
Melting Pt: 174…176 °C.
MDL Number: MFCD00052268
CAS Number: 76195-82-9

Order Now

Specification Test Results

Melting point 185°(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR