3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane ≥97%

Supplier: Matrix Scientific

Synonyms: [3,5-Bis(trifluoromethyl)phenyl]chloro(dimethyl)silane

023808-1G 023808-5G
101780-348EA 124.62 USD
101780-348 101780-350
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane ≥97%
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane

Hazard: Corrosive/Moisture Sensitive

Formula: C₁₀H₉ClF₆Si
MW: 306.71 g/mol
Boiling Pt: 95…96 °C (30 mmHg)
MDL Number: MFCD01862186
CAS Number: 732306-23-9

Order Now

Specification Test Results

Boiling point 95-96°/30m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR