4-Bromo-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-(trifluoromethyl)benzene-1-sulphonyl chloride

023760-5G 023760-25G
101780-338EA 33.85 USD
101780-338 101780-340
4-Bromo-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive/Irritant

Formula: C₇H₃BrClF₃O₂S
MW: 323.52 g/mol
Melting Pt: 54…58 °C
MDL Number: MFCD03094392
CAS Number: 176225-10-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 54-58°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR