3-Fluoro-4-methylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methylbenzene-1-sulphonyl chloride

023751-5G 023751-25G
101780-334EA 35.38 USD
101780-334 101780-336
3-Fluoro-4-methylbenzenesulfonyl chloride ≥97%
3-Fluoro-4-methylbenzenesulphonyl chloride

Hazard: Corrosive/Irritant

Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
MDL Number: MFCD03094389
CAS Number: 90260-13-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR