2-Bromothiazole ≥97%

Supplier: Matrix Scientific

023743-25G 023743-100G
101780-330EA 18.46 USD
101780-330 101780-332
2-Bromothiazole ≥97%
2-Bromothiazole

Hazard: Flammable/Irritant/Refrigerate

Formula: C₃H₂BrNS
Boiling Pt: 70…71 °C (20 mmHg)
Density: 1.836
MDL Number: MFCD00005316
CAS Number: 3034-53-5

Order Now

Specification Test Results

Boiling point 70-71°/20m
Density 1.836


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR