2-Bromo-4,6-difluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

023740-25G 023740-5G
101780-328EA 129.23 USD
101780-328 101780-326
2-Bromo-4,6-difluorobenzenesulfonyl chloride ≥97%
2-Bromo-4,6-difluorobenzenesulphonyl chloride

Hazard: Corrosive/Irritant

Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Melting Pt: 41…44 °C
MDL Number: MFCD03094561
CAS Number: 351003-42-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Melting point 41-44°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR