2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(trifluoromethyl)benzene-1-sulphonyl chloride

023738-1G 023738-5G
101780-322EA 29.23 USD
101780-322 101780-324
2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride ≥97%
2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride

Hazard: Flammable/Corrosive/Moisture Sensitive

Formula: C₇H₃BrClF₃O₂S
MW: 323.52 g/mol
Boiling Pt: 235-236°C
Melting Pt: 36-39°C
Density: 1.865
MDL Number: MFCD03094394
CAS Number: 54403-98-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 235-236°
Density 1.865
ND20 1.527

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR