2-Chloropyridine-5-acetonitrile ≥98%

Supplier: Matrix Scientific

023735-25G 023735-100G
101780-320EA 220 USD
101780-320 102832-914
2-Chloropyridine-5-acetonitrile ≥98%
2-Chloropyridine-5-acetonitrile

Hazard: Toxic/Irritant

Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
MDL Number: MFCD04112495
CAS Number: 39891-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR