Chloromethanesulfonyl chloride ≥95%
Supplier: Matrix Scientific
023718-5G
023718-25G
101780-314EA
146.15
USD
101780-314
101780-316
Chloromethanesulfonyl chloride ≥95%
Chloromethanesulfonyl chloride
Hazard: Toxic/Corrosive/Refrigerate
Formula:
CH₂Cl₂O₂S Boiling Pt: 75°C Density: 1.638 |
MDL Number:
MFCD00014727 CAS Number: 3518-65-8 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 80-81°/25m |
Density | 1.67 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...