2-Chloro-3-methyl-5-nitropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-nitro-3-picoline

021401-5G 021401-25G
101780-310EA 16.92 USD
101780-310 101780-312
2-Chloro-3-methyl-5-nitropyridine ≥98%
2-Chloro-3-methyl-5-nitropyridine

Hazard: Toxic/Irritant

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD00173686
CAS Number: 22280-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR