5-Amino-2-(methylthio)benzotrifluoride ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Methylthio-3-(trifluoromethyl)aniline
021360-1G
021360-5G
101780-306EA
47.69
USD
101780-306
101780-308
5-Amino-2-(methylthio)benzotrifluoride ≥98%
5-Amino-2-(methylthio)benzotrifluoride
Hazard: Toxic
Formula:
C₈H₈F₃NS MW: 207.22 g/mol Melting Pt: 58…60 °C |
MDL Number:
MFCD04972923 CAS Number: 63094-56-4 UN: 2811 ADR: 6.1 III |
Specification Test Results
Melting point | 58-60° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...