4-Amino-2-methylpyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine
021150-5G
021150-25G
021150-100G
101780-304EA
15.38
USD
101780-304
102551-008
102551-010
4-Amino-2-methylpyridine ≥98%
4-Amino-2-methylpyridine
Hazard: Toxic/Irritant
Formula:
C₆H₈N₂ MW: 108.14 g/mol Melting Pt: 93…97 °C |
MDL Number:
MFCD00186509 CAS Number: 18437-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...