4-Amino-3-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

021130-5G 021130-25G
101780-290EA 23.08 USD
101780-290 101780-292
4-Amino-3-fluorobenzonitrile ≥98%
4-Amino-3-fluorobenzonitrile

Hazard: Toxic/Irritant

Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD00055559
CAS Number: 63069-50-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR