2,3-Difluoropyridine ≥98%

Supplier: Matrix Scientific

021119-25G 021119-1G
101780-284EA 164.62 USD
101780-284 101780-280
2,3-Difluoropyridine ≥98%
2,3-Difluoropyridine

Hazard: Flammable

Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 125-126°C
MDL Number: MFCD04114143
CAS Number: 1513-66-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 125-126°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR