5-Fluoro-o-toluonitrile ≥98%

Supplier: Matrix Scientific

021043-25G 021043-100G
101780-272EA 49.23 USD
101780-272 101780-274
5-Fluoro-o-toluonitrile ≥98%

Hazard: Toxic

Formula: C₈H₆FN
Melting Pt: 42…44 °C
MDL Number: MFCD00042295
CAS Number: 77532-79-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 42-44°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR