4-Chloro-2,5-dimethylbenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2,5-dimethylbenzene-1-sulphonyl chloride, 5-Chloro-p-xylene-2-sulphonyl chloride

020745-25G 020745-5G
101780-262EA 175.38 USD
101780-262 101780-260
4-Chloro-2,5-dimethylbenzenesulfonyl chloride ≥98%
4-Chloro-2,5-dimethylbenzenesulphonyl chloride

Hazard: Corrosive/Water Reactive

Formula: C₈H₈Cl₂O₂S
MW: 239.12 g/mol
Melting Pt: 48-50°C
MDL Number: MFCD00044017
CAS Number: 88-49-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 48-50°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR