2-Chloro-5-(chloromethyl)pyridine ≥95%

Supplier: Matrix Scientific

020606-25G 020606-100G
101780-252EA 20 USD
101780-252 101780-254
2-Chloro-5-(chloromethyl)pyridine ≥95%
2-Chloro-5-(chloromethyl)pyridine

Hazard: Corrosive/Lachrymator

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 37…42 °C
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 37-42°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR