Ethyl-4,4,4-trifluorobut-2-ynoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,4,4-trifluorobut-2-ynoate

020176-500MG 020176-2.5G
101780-238EA 112.31 USD
101780-238 101780-240
Ethyl-4,4,4-trifluorobut-2-ynoate ≥97%
Ethyl-4,4,4-trifluorobut-2-ynoate

Hazard: Flammable/Refrigerate/Lachrymator

Formula: C₆H₅F₃O₂
MW: 166.099 g/mol
Boiling Pt: 96-98°C
Density: 1.162
Flash Pt: 6°C
MDL Number: MFCD00192167
CAS Number: 79424-03-6

Order Now

Specification Test Results

Boiling point 96-98°
Density 1.162
ND20 1.351

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR