2-Methyl-5-nitrobenzenesulphonyl chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Nitrotoluene-2-sulphonyl chloride, 2-Methyl-5-nitrobenzene-1-sulphonyl chloride
018319-5G
018319-25G
101780-224EA
29.23
USD
101780-224
101780-226
2-Methyl-5-nitrobenzenesulphonyl chloride ≥95%
2-Methyl-5-nitrobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₇H₆ClNO₄S Melting Pt: 43…45 °C |
MDL Number:
MFCD00051695 CAS Number: 121-02-8 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 43-45° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...