3-Bromo-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

018268-1G 018268-5G
101780-204EA 21.54 USD
101780-204 101780-206
3-Bromo-5-(trifluoromethyl)pyridine ≥97%
3-Bromo-5-(trifluoromethyl)pyridine

Hazard: Toxic

Formula: C₆H₃BrF₃N
MW: 226.00 g/mol
Boiling Pt: 43°C/14mm
Melting Pt: 35-36°C
MDL Number: MFCD04972700
CAS Number: 436799-33-6

Order Now

Specification Test Results

Boiling point 43°/14mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR