3-Bromo-2-methyl-1-propene ≥95%

Supplier: Matrix Scientific

018253-5G 018253-25G 018253-100G
101780-194EA 36.92 USD
101780-194 101780-196 101780-198
3-Bromo-2-methyl-1-propene ≥95%
3-Bromo-2-methyl-1-propene

Hazard: Flammable/Refrigerate/Lachrymator

Formula: C₄H₇Br
Boiling Pt: 94-95°C
Density: 1.339
MDL Number: MFCD00134155
CAS Number: 1458-98-6

Order Now

Specification Test Results

Boiling point 94-95°
Density 1.339
ND20 14720


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR