Trifluorolactic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoro-2-hydroxypropanoic acid

018209-1G 018209-5G
101780-190EA 121.54 USD
101780-190 101780-192
Trifluorolactic acid ≥98%
Trifluorolactic acid

Hazard: Corrosive/Hygroscopic

Formula: C₃H₃F₃O₃
MW: 144.05 g/mol
Boiling Pt: 157 °C (123 mmHg)
Melting Pt: 66…70 °C
MDL Number: MFCD00051802
CAS Number: 684-07-1

Order Now

Specification Test Results

Boiling point 157°/123mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR