4-n-Propylpiperidine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Propylpiperidine

016551-250MG 016551-1G
101780-184EA 73.85 USD
101780-184 101780-186
4-n-Propylpiperidine ≥97%
4-n-Propylpiperidine

Hazard: Flammable

Formula: C₈H₁₇N
MW: 127.23 g/mol
MDL Number: MFCD00023151
CAS Number: 22398-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR