α,3,5-Tribromotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
016548-5G
016548-25G
101780-180EA
89.23
USD
101780-180
101780-182
α,3,5-Tribromotoluene ≥98%
α,3,5-Tribromotoluene
Hazard: Corrosive/Lachrymator
Formula:
C₇H₅Br₃ MW: 328.83 g/mol Melting Pt: 93…95 °C |
MDL Number:
MFCD00052415 CAS Number: 56908-88-4 UN: 1759 ADR: 8,III |
Specification Test Results
Melting point | 93-95° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...