2,4-Dibromo-6-fluoroaniline ≥98%

Supplier: Matrix Scientific

016045-100G 016045-25G
101780-178EA 160 USD
101780-178 101780-176
2,4-Dibromo-6-fluoroaniline ≥98%
2,4-Dibromo-6-fluoroaniline

Hazard: Toxic/Irritant

Formula: C₆H₄Br₂FN
MW: 268.91 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD00042230
CAS Number: 141474-37-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 62-64°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR