(Trifluoromethyl)maleic anhydride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)furan-2,5-dione

012896-1G 012896-5G
101780-168EA 84.62 USD
101780-168 101780-170
(Trifluoromethyl)maleic anhydride ≥97%
(Trifluoromethyl)maleic anhydride

Hazard: Corrosive/Irritant

Formula: C₅HF₃O₃
MW: 166.06 g/mol
Boiling Pt: 93 °C (65 mmHg) Hg
Flash Pt: 37 °C
MDL Number: MFCD00153287
CAS Number: 700-27-6

Order Now

Specification Test Results

Boiling point 93°/65mm H

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR