2,4-Dichloro-5-fluoropyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2,4-dichloropyrimidine

012656-100G 012656-25G
101780-148EA 75.38 USD
101780-148 101780-146
2,4-Dichloro-5-fluoropyrimidine ≥98%
2,4-Dichloro-5-fluoropyrimidine

Hazard: Corrosive/Avoid Skin Contact/Lachrymator

Formula: C₄HCl₂FN₂
MW: 166.97 g/mol
Boiling Pt: 74…78 °C (16 Torr)
Melting Pt: 37…41 °C
Flash Pt: 106 °C
MDL Number: MFCD00233551
CAS Number: 2927-71-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR