3,3,3-Trifluoropropane-1-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoropropane-1-sulphonylchloride

012616-1G 012616-5G
101780-134EA 126.15 USD
101780-134 101780-136
3,3,3-Trifluoropropane-1-sulfonyl chloride ≥95%
3,3,3-Trifluoropropane-1-sulfonyl chloride

Hazard: Corrosive/Irritant

Formula: C₃H₄ClF₃O₂S
MW: 196.58 g/mol
Boiling Pt: 76…78 °C (16 mmHg)
MDL Number: MFCD06248898
CAS Number: 845866-80-0

Order Now

Specification Test Results

Boiling point 76-78°/16m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR