4-Amino-1-boc-piperidine ≥98%

Supplier: Matrix Scientific

012595-25G 012595-100G
101780-122EA 44.62 USD
101780-122 101780-124
4-Amino-1-boc-piperidine ≥98%
4-Amino-1-Boc-piperidine

Hazard: Corrosive/Air Sensitive

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 80 °C (0.05 mmHg)
Melting Pt: 44…48 °C
MDL Number: MFCD01076201
CAS Number: 87120-72-7

Order Now

Specification Test Results

Boiling point 80°/0.05mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR