4-Aminotetrahydropyran ≥98%

Supplier: Matrix Scientific

Synonyms: Tetrahydro-2H-pyran-4-amine

012574-1G 012574-5G
101780-118EA 23.08 USD
101780-118 101780-120
4-Aminotetrahydropyran ≥98%
4-Aminotetrahydropyran

Hazard: Flammable/Stored under Argon

Formula: C₅H₁₁NO
MW: 101.1469 g/mol
Boiling Pt: 60°C/12mm
MDL Number: MFCD02179436
CAS Number: 38041-19-9

Order Now

Specification Test Results

Boiling point 60°/12mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR